KEY456787255 Key Organics / BIONET

potassium (3-ethoxyphenyl)trifluoroboranuide

  • MDL number MFCD09993311
  • Cas Number : 1638533-83-1
  • Molecular Weight : 228.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

signalword Warning
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart