KEY456787258 Key Organics / BIONET

1-benzyl-2-methylbenzene

  • MDL number MFCD00270116
  • Cas Number : 713-36-0
  • Molecular Weight : 182.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/519653

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3168_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart