KEY458250908 Key Organics / BIONET

5-nitro-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD00232024
  • Cas Number : 82827-08-5
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/10B-099_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart