KEY458250981 Key Organics / BIONET

methyl 3-aminothieno[2,3-c]pyridine-2-carboxylate

  • MDL number MFCD04124001
  • Cas Number : 111042-91-2
  • Molecular Weight : 208.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-aminothieno[2,3-c]pyridine-2-carboxylate
Inchi Code 1S/C9H8N2O2S/c1-13-9(12)8-7(10)5-2-3-11-4-6(5)14-8/h2-4H,10H2,1H3
Inchi Key VAAFWMASRDVMQF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/202

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/10D-065_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart