KEY458251000 Key Organics / BIONET

ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate

  • MDL number MFCD00139509
  • Cas Number : 2549-19-1
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate
Inchi Code 1S/C11H12N2O2/c1-3-15-11(14)10-8(2)12-9-6-4-5-7-13(9)10/h4-7H,3H2,1-2H3
Inchi Key FLAQFBICEAEEOA-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/239

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/10D-518_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart