KEY458251061 Key Organics / BIONET

1-benzyl-5-(4-ethyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)-1,2-dihydropyridin-2-one

  • MDL number MFCD00140624
  • Cas Number : 242472-12-4
  • Molecular Weight : 312.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/371

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/10E-390S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart