KEY458251185 Key Organics / BIONET

2-chloro-6-(methylsulfanyl)pyridine

  • MDL number MFCD00173422
  • Cas Number : 77145-64-3
  • Molecular Weight : 159.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-6-(methylsulfanyl)pyridine
Inchi Code 1S/C6H6ClNS/c1-9-6-4-2-3-5(7)8-6/h2-4H,1H3
Inchi Key KVYNCXZSIHYSDF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/654

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/10G-006_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart