KEY458251235 Key Organics / BIONET

7-chloroquinazolin-4-ol

  • MDL number MFCD00115130
  • Cas Number : 31374-18-2
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloro-4(3H)-quinazolinone
Inchi Code 1S/C8H5ClN2O/c9-5-1-2-6-7(3-5)10-4-11-8(6)12/h1-4H,(H,10,11,12)
Inchi Key PMLONMIODRHERC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2389

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/10N-832_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart