KEY458251249 Key Organics / BIONET

(3-methylpyridin-2-yl)thiourea

  • MDL number MFCD03617970
  • Cas Number : 41440-07-7
  • Molecular Weight : 167.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3-methyl-2-pyridinyl)thiourea
Inchi Code 1S/C7H9N3S/c1-5-3-2-4-9-6(5)10-7(8)11/h2-4H,1H3,(H3,8,9,10,11)
Inchi Key GHHXGWZMJBHPOM-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2664

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/10P-930_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart