KEY458251270 Key Organics / BIONET

2-(1-methylhydrazin-1-yl)pyrazine

  • MDL number MFCD05975001
  • Cas Number : 76319-95-4
  • Molecular Weight : 124.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1-methylhydrazino)pyrazine
Inchi Code 1S/C5H8N4/c1-9(6)5-4-7-2-3-8-5/h2-4H,6H2,1H3
Inchi Key ZINDLWVBIZZAOY-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3543

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/10X-0840_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart