KEY458251279 Key Organics / BIONET

ethyl[(pyridin-3-yl)methyl]amine

  • MDL number MFCD04557869
  • Cas Number : 3000-75-7
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3-pyridinylmethyl)ethanamine
Inchi Code 1S/C8H12N2/c1-2-9-6-8-4-3-5-10-7-8/h3-5,7,9H,2,6H2,1H3
Inchi Key SGROJTFSHJVVSF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3709

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/10Z-5027_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart