KEY458251283 Key Organics / BIONET

(2E)-3-(dimethylamino)-1-(4-methylphenyl)prop-2-en-1-one

  • MDL number MFCD00121190
  • Cas Number : 502841-03-4
  • Molecular Weight : 189.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3848

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11C-009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart