KEY458251288 Key Organics / BIONET

1-[(3-chlorophenyl)methyl]-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00141641
  • Cas Number : 206537-57-7
  • Molecular Weight : 271.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4313

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-375S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart