KEY458251291 Key Organics / BIONET

3-oxo-N-(1,3-thiazol-2-yl)butanamide

  • MDL number MFCD00022444
  • Cas Number : 705-87-3
  • Molecular Weight : 184.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4425

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-954_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart