KEY458251293 Key Organics / BIONET

N-[2-(1H-pyrrol-1-yl)ethyl]acetamide

  • MDL number MFCD00180783
  • Cas Number : 73627-16-4
  • Molecular Weight : 152.197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4618

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/11G-936_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart