KEY458251297 Key Organics / BIONET

1-(1H-1,3-benzodiazol-2-yl)ethan-1-ol

  • MDL number MFCD00016718
  • Cas Number : 19018-24-7
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(1H-benzimidazol-2-yl)ethanol
Inchi Code 1S/C9H10N2O/c1-6(12)9-10-7-4-2-3-5-8(7)11-9/h2-6,12H,1H3,(H,10,11)
Inchi Key XZHWEHOSQYNGOL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4677

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11H-051_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart