KEY458251332 Key Organics / BIONET

5-(4-tert-butylphenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD00500939
  • Cas Number : 306936-90-3
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(4-tert-butylphenyl)-1,3,4-oxadiazole-2(3H)-thione
Inchi Code 1S/C12H14N2OS/c1-12(2,3)9-6-4-8(5-7-9)10-13-14-11(16)15-10/h4-7H,1-3H3,(H,14,16)
Inchi Key ATIYHEZPBLMRNI-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6313

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-179_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart