KEY458251340 Key Organics / BIONET

2,5-dimethylpyrazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD00995337
  • Cas Number : 2369-89-3
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-dimethylpyrazolo[1,5-a]pyrimidin-7-amine
Inchi Code 1S/C8H10N4/c1-5-3-7(9)12-8(10-5)4-6(2)11-12/h3-4,10H,1,9H2,2H3
Inchi Key LZQISAIVDKZPOR-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6637

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0075_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart