KEY458251349 Key Organics / BIONET

1-(2-methyl-3-nitrophenyl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD02664862
  • Cas Number : 383136-10-5
  • Molecular Weight : 230.223

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7064

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0847_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart