KEY458251355 Key Organics / BIONET

(5Z)-2-sulfanylidene-5-{[2-(trifluoromethyl)phenyl]methylidene}-1,3-thiazolidin-4-one

  • MDL number MFCD01018636
  • Cas Number : 917985-96-7
  • Molecular Weight : 289.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7129

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0247_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart