KEY458251369 Key Organics / BIONET

1-[(2,4-dichlorophenyl)methyl]-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00141578
  • Cas Number : 79183-24-7
  • Molecular Weight : 306.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,4-dichlorobenzyl)-1H-indole-2,3-dione
Inchi Code 1S/C15H9Cl2NO2/c16-10-6-5-9(12(17)7-10)8-18-13-4-2-1-3-11(13)14(19)15(18)20/h1-7H,8H2
Inchi Key KRLHWKJGYCSOLQ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7708

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12E-324S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart