KEY458251373 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperidin-4-ol

  • MDL number MFCD00202495
  • Cas Number : 306976-40-9
  • Molecular Weight : 280.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinol
Inchi Code 1S/C11H12ClF3N2O/c12-9-5-7(11(13,14)15)6-16-10(9)17-3-1-8(18)2-4-17/h5-6,8,18H,1-4H2
Inchi Key QFDHQYSCNBOECL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7972

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12F-951_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart