KEY458251377 Key Organics / BIONET

ethyl N-[(5-chloropyridin-2-yl)carbamothioyl]carbamate

  • MDL number MFCD01833204
  • Cas Number : 78852-50-3
  • Molecular Weight : 259.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl [(5-chloro-2-pyridinyl)amino]carbothioylcarbamate
Inchi Code 1S/C9H10ClN3O2S/c1-2-15-9(14)13-8(16)12-7-4-3-6(10)5-11-7/h3-5H,2H2,1H3,(H2,11,12,13,14,16)
Inchi Key SUDFDUAHBQUCKX-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8341

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12J-530S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart