KEY458251380 Key Organics / BIONET

[(E)-[(4-methoxyphenyl)methylidene]amino]urea

  • MDL number MFCD00456389
  • Cas Number : 120445-53-6
  • Molecular Weight : 193.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209 - 210
Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8774

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart