KEY458251385 Key Organics / BIONET

4-(4-fluorophenoxy)benzonitrile

  • MDL number MFCD01815090
  • Cas Number : 215589-24-5
  • Molecular Weight : 213.211

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9035

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-083_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart