KEY458251388 Key Organics / BIONET

2-bromo-6-butoxynaphthalene

  • MDL number MFCD01568860
  • Cas Number : 66217-20-7
  • Molecular Weight : 279.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-6-butoxynaphthalene
Inchi Code 1S/C14H15BrO/c1-2-3-8-16-14-7-5-11-9-13(15)6-4-12(11)10-14/h4-7,9-10H,2-3,8H2,1H3
Inchi Key LIQQRPRDWMYCPK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9045

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-111_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart