KEY458251402 Key Organics / BIONET

1-(benzyloxy)-4-ethynylbenzene

  • MDL number MFCD01814885
  • Cas Number : 84284-70-8
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 4-ethynylphenyl ether
Inchi Code 1S/C15H12O/c1-2-13-8-10-15(11-9-13)16-12-14-6-4-3-5-7-14/h1,3-11H,12H2
Inchi Key XQHAOXRZNLCKJO-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9120

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-249_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart