KEY458251408 Key Organics / BIONET

4-(1-hydroxypropan-2-yl)-1lambda6-thiomorpholine-1,1-dione

  • MDL number MFCD02186066
  • Cas Number : 477864-26-9
  • Molecular Weight : 193.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9533

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12P-622_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart