KEY458251410 Key Organics / BIONET

3-(4-methylpiperazin-1-yl)-3-oxopropanenitrile

  • MDL number MFCD02186990
  • Cas Number : 15029-34-2
  • Molecular Weight : 167.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9559

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-665_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart