KEY458251414 Key Organics / BIONET

ethyl 4-chloro-1-ethyl-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD04124526
  • Cas Number : 37801-47-1
  • Molecular Weight : 267.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9936

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12W-0020_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart