KEY458251415 Key Organics / BIONET

4-chloro-N-ethyl-3-nitro-N-phenylbenzene-1-sulfonamide

  • MDL number MFCD02724844
  • Cas Number : 380335-72-8
  • Molecular Weight : 340.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9937

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart