KEY458251416 Key Organics / BIONET

2-chloro-N-ethyl-N-phenylacetamide

  • MDL number MFCD00227876
  • Cas Number : 39086-61-8
  • Molecular Weight : 197.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9946

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12W-0045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart