KEY458251432 Key Organics / BIONET

methyl 4-amino-3-bromo-5-methylbenzoate

  • MDL number MFCD07021337
  • Cas Number : 900019-52-5
  • Molecular Weight : 244.088

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63038

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12X-0818_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart