KEY458251433 Key Organics / BIONET

4-amino-3-bromo-5-methylbenzoic acid

  • MDL number MFCD05975057
  • Cas Number : 860787-42-4
  • Molecular Weight : 230.061

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63041

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12X-0821_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart