KEY458251442 Key Organics / BIONET

(4Z)-4-[(dimethylamino)methylidene]-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00100702
  • Cas Number : 65644-13-5
  • Molecular Weight : 229.283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63190

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1C-045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart