KEY458251444 Key Organics / BIONET

1-{7-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-6-yl}ethan-1-one

  • MDL number MFCD00244648
  • Cas Number : 320416-90-8
  • Molecular Weight : 176.179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63335

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1D-095_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart