KEY458251446 Key Organics / BIONET

7-amino-6-phenylpyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD00138891
  • Cas Number : 320417-39-8
  • Molecular Weight : 235.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63371

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1D-153_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart