KEY458251451 Key Organics / BIONET

N-(2,4-dichlorophenyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD00751435
  • Cas Number : 121806-47-1
  • Molecular Weight : 258.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64276

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1J-944_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart