KEY458251457 Key Organics / BIONET

4H,5H-pyrrolo[1,2-a]quinoxalin-4-one

  • MDL number MFCD00275577
  • Cas Number : 6025-68-9
  • Molecular Weight : 184.198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64921

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1N-004_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart