KEY458251461 Key Organics / BIONET

methyl 2-(2,2,2-trifluoroacetamido)benzoate

  • MDL number MFCD00436637
  • Cas Number : 31385-12-3
  • Molecular Weight : 247.173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64984

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1N-543S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart