KEY458251462 Key Organics / BIONET

ethyl 2-(morpholin-4-yl)-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD01315302
  • Cas Number : 338392-97-5
  • Molecular Weight : 310.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64990

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1N-557S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart