KEY458251468 Key Organics / BIONET

2-{[(4-fluorophenyl)methyl]amino}acetonitrile

  • MDL number MFCD02102395
  • Cas Number : 63086-22-6
  • Molecular Weight : 164.183

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65431

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0185_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart