KEY458251473 Key Organics / BIONET

5-chloro-6-phenyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD00218559
  • Cas Number : 51660-08-3
  • Molecular Weight : 206.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65614

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-1068_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart