KEY458251476 Key Organics / BIONET

2-[(4-methoxyphenyl)methyl]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD02102515
  • Cas Number : 477872-94-9
  • Molecular Weight : 249.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65717

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-1536_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart