KEY458251477 Key Organics / BIONET

ethyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate

  • MDL number MFCD01956277
  • Cas Number : 82791-95-5
  • Molecular Weight : 272.227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65719

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1543_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart