KEY458251481 Key Organics / BIONET

1,5-dimethyl-4-phenylimidazolidin-2-one

  • MDL number MFCD01456559
  • Cas Number : 103774-40-9
  • Molecular Weight : 190.246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65790

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0277_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart