KEY458251482 Key Organics / BIONET

1-(4-bromophenyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD03011828
  • Cas Number : 383148-16-1
  • Molecular Weight : 266.094

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65825

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0601_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart