KEY458251487 Key Organics / BIONET

2-[(6-chloropyridin-3-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD09802551
  • Cas Number : 120739-60-8
  • Molecular Weight : 272.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66016

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1W-0205_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart