KEY458251503 Key Organics / BIONET

2-({5-methylthieno[2,3-d]pyrimidin-4-yl}sulfanyl)aniline

  • MDL number MFCD00110570
  • Cas Number : 175137-24-3
  • Molecular Weight : 273.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66450

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0929_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart