KEY458251508 Key Organics / BIONET

N-[(furan-2-yl)methyl]-2-nitrobenzene-1-sulfonamide

  • MDL number MFCD00686900
  • Cas Number : 303063-01-6
  • Molecular Weight : 282.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66551

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0946_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart